missing translation for 'onlineSavingsMsg'
Learn More

Restek™ Rinse/Waste Vials with Diffusion Caps

Catalog No. 06611261
Encompass
Change view
Click to view available options
Color:
Clear
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
06611261 Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06611261 Supplier Restek™ Supplier No. 23096

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Diffusion caps prevent evaporation of volatile solvents

  • Six different cap colors for easy solvent identification
  • Graduated marks on vials indicate content volume
  • Includes twelve vials and caps (two caps of each color)

Specifications

Closure Color Assorted
Color Clear
Material Glass
Pre-slit No
Sterility Non-sterile
Disposable Single-use
Volume (Metric) 4 mL
Graduated No
Style Rinse/Waste
Closure Type Diffusion
Quantity 12 Ea.
Marking Spot Yes
Preservative No
Show More Show Less