missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rinse Solution, High Purity Water for Rinsing Electrodes and other Laboratory Instruments, Ricca Chemical
$64.31 - $64.31
Chemical Identifiers
| CAS | 7732-18-5 |
|---|---|
| Molecular Formula | H2O |
| Molecular Weight (g/mol) | 18.015 |
| InChI Key | XLYOFNOQVPJJNP-UHFFFAOYSA-N |
| Synonym | dihydrogen oxide, dihydrogen monoxide |
| PubChem CID | 962 |
| ChEBI | CHEBI:15377 |
| IUPAC Name | oxidane |
| SMILES | O |
Chemical Identifiers
| 7732-18-5 | |
| 18.015 | |
| dihydrogen oxide, dihydrogen monoxide | |
| CHEBI:15377 | |
| O |
| H2O | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| 962 | |
| oxidane |
Specifications
| Colorless | |
| 98.0 | |
| H2O | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| oxidane | |
| 962 | |
| 100% | |
| Natural Poly Bottle | |
| Liquid |
| 7732-18-5 | |
| 100.0 | |
| dihydrogen oxide, dihydrogen monoxide | |
| O | |
| 18.015 | |
| CHEBI:15377 | |
| Laboratory | |
| 500 mL | |
| Rinse Solution, High Purity Water for Rinsing Electrodes and other Laboratory Instruments |