missing translation for 'onlineSavingsMsg'
Learn More

Riluzole, 98%

Catalog No. AC459360050
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC459360050 5 g
AC459360010 1 g
AC459360100 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC459360050 Supplier Thermo Scientific Chemicals Supplier No. 459360050
Only null left

CAS: 1744-22-5 | C8H5F3N2OS | 234.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1744-22-5
Molecular Formula C8H5F3N2OS
Molecular Weight (g/mol) 234.2
InChI Key FTALBRSUTCGOEG-UHFFFAOYSA-N
Synonym riluzole, rilutek, 2-amino-6-trifluoromethoxy benzothiazole, 6-trifluoromethoxy benzo d thiazol-2-amine, 6-trifluoromethoxy-1,3-benzothiazol-2-amine, riluzolum, riluzol, 2-amino-6-trifluoromethoxybenzothiazole, riluzole hcl, riluzol inn-spanish
PubChem CID 5070
ChEBI CHEBI:8863
IUPAC Name 6-(trifluoromethoxy)-1,3-benzothiazol-2-amine
SMILES C1=CC2=C(C=C1OC(F)(F)F)SC(=N2)N

Specifications

Melting Point 114.0°C to 118.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 5 g
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in methanol,ether,acetone,dmf,chlorofo,rm
Formula Weight 234.2
Percent Purity 98%
Chemical Name or Material Riluzole
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Fatal if swallowed.
Precautionary Statement GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Rinse mouth.

RUO – Research Use Only