accessibility menu, dialog, popup

UserName

CAS RN 1744-22-5

IUPAC Name: 6-(trifluoromethoxy)-1,3-benzothiazol-2-amine
Synonyms: riluzole rilutek 利鲁唑
Molecular Weight (g/mol): 234.2
Molecular Formula: C8H5F3N2OS
InChi Key: FTALBRSUTCGOEG-UHFFFAOYSA-N
SMILES: NC1=NC2=CC=C(OC(F)(F)F)C=C2S1
Molecular Weight (g/mol) 
  • (4)
  • (3)
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (1)
  • (1)
  • (2)
  • (3)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (4)
  • (3)
  • (2)
  • (5)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)