missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Right-Angled Glass Prisms

Non-optically worked with polished faces.

Supplier:  Eisco™ PH0558B

 View more versions of this product

Catalog No. S14033


Only null left
Explore available promotions

Description

Description

  • Edges slightly bevelled
  • Nominal angles of 90° x 45° x 45°
Specifications

Specifications

Prism, Right-angled Glass
Nominal angles of 90° x 45° x 45°
11.4 x 9.4 x 2 in.
35 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions