Learn More
Eisco™ Right-Angled Glass Prisms
Catalog No. p-6962034
Change view
Click to view available options
Length (Metric):
25 mm
35 mm
50 mm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
S14032 25 mm
S14033 35 mm
S14034 50 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S14032 Supplier Eisco™ Supplier No. PH0558A
Only null left
Non-optically worked with polished faces.
  • With polished faces
  • Edges slightly bevelled
  • Nominal angles of 90 x 45 x 45 deg.
Specifications
Product Type Prism, Right-angled Glass
Description Nominal angles of 90 x 45 x 45 deg.
Length (Metric) 25 mm