Learn More
Eisco™ Right-Angled Glass Prisms
Non-optically worked with polished faces.
Supplier: Eisco™ PH0558A
Description
- With polished faces
- Edges slightly bevelled
- Nominal angles of 90 x 45 x 45 deg.
Specifications
| Prism, Right-angled Glass | |
| Nominal angles of 90 x 45 x 45 deg. | |
| 25 mm |