missing translation for 'onlineSavingsMsg'
Learn More

Rifamycin Sodium Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111013176
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013176 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013176 Supplier Merck Emd Millipore Supplier No. PHR1664500MG
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Rifamycin Sodium is a simplest rifamycin obtained via the elimination of glycolic moity from rifamycin B. It belongs to the ansamicin group of antibiotics. Rifamycin sodium is widely used in surgery and for the treatment of synovitis by intracellular injections.

Specifications

CAS 14897-39-3
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C37H46 NNaO12
Synonym Rifamycin SV sodium salt
Type Certified Reference Material
Quantity 500 mg
Formula Weight 719.75
Grade Certified Reference Material