missing translation for 'onlineSavingsMsg'
Learn More

Rifampin Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111012171
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012171 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012171 Supplier Merck Emd Millipore Supplier No. PHR1806500MG

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Specifications

CAS 13292-46-1
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C43H58N4O12
MDL Number MFCD00151389
Synonym Rifampicin; 3-(4-Methylpiperazinyliminomethyl)rifamycin SV; Rifampin; Rifamycin AMP
Type Certified Reference Material
Quantity 500 mg
Formula Weight 822.94
Grade Certified Reference Material