Learn More
Epredia™ Decalcifying Solution
Remove calcium from histological specimens with Epredia™ Decalcifying Solution.
$233.64 - $395.71
Specifications
Product Type Decalcifying Solution
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
361014333
View Documents Promotion Details
Epredia™
8340
32 oz.
Case of 6 Each for $395.71
Only null left
22050263
View Documents Promotion Details
Epredia™
83401
1 gal.
Each for $233.64
Case of 4 Each for $934.56
Only null left
Description
Description
  • Offers rapid decalcification without destroying nuclear properties of the tissue
  • The EDTA and dilute hydrochloric acid in this solution require no heat, electricity, or other aids to decalcify bone tissue
Specifications
Specifications
Decalcifying Solution