Learn More
Epredia™ Decalcifying Solution
Catalog No. 361014333
Change view
Click to view available options
Quantity:
32 oz.
1 gal.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
361014333 32 oz.
22050263 1 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 361014333 Supplier Epredia™ Supplier No. 8340
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Remove calcium from histological specimens with Epredia™ Decalcifying Solution.
  • Offers rapid decalcification without destroying nuclear properties of the tissue
  • The EDTA and dilute hydrochloric acid in this solution require no heat, electricity, or other aids to decalcify bone tissue
Specifications
Product Type Decalcifying Solution
Quantity 32 oz.