missing translation for 'onlineSavingsMsg'
Learn More
Learn More
pH Test Strips, Ricca Chemical
General-purpose pH test strips
Supplier: Ricca Chemical Company R8882000EACH
Description
- Available in four different pH ranges
- Time-saving, single-use indicator strips with color-fixed test area on an easy-to-handle, durable plastic strip
- Fast accurate results
- Packages include a color comparison chart
- 100 strips per pack
Chemical Identifiers
| For precise pH determination | |
| Color chart | |
| pH Test Strip |
Specifications
| 4.5 to 10 | |
| 0.5 | |
| 100 Strips |
| For precise pH determination | |
| Color chart | |
| pH Test Strip |