missing translation for 'onlineSavingsMsg'
Learn More

Ribostamycin Sulfate 90.0+%, TCI America™

Catalog No. r01941g
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R01941G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R01941G Supplier TCI America Supplier No. R01941G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 53797-35-6
Molecular Formula C17H36N4O14S
Molecular Weight (g/mol) 552.55
MDL Number MFCD23380549
InChI Key RTCDDYYZMGGHOE-YMSVYGIHSA-N
PubChem CID 53486170
IUPAC Name (2R,3S,4R,5R,6R)-5-amino-2-(aminomethyl)-6-{[(1R,2R,3S,4R,6S)-4,6-diamino-2-{[(2S,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]oxy}-3-hydroxycyclohexyl]oxy}oxane-3,4-diol; sulfuric acid
SMILES OS(O)(=O)=O.NC[C@H]1O[C@H](O[C@@H]2[C@@H](N)C[C@@H](N)[C@H](O)[C@H]2O[C@@H]2O[C@H](CO)[C@@H](O)[C@H]2O)[C@H](N)[C@@H](O)[C@@H]1O

Specifications

Type Ribostamycin Sulfate
Physical Form Crystalline Powder
Melting Point 180°C
Color White
Quantity 1 g
Percent Purity ≥90.0% (N)
Chemical Name or Material Ribostamycin Sulfate
TSCA No
Recommended Storage Refrigerator