accessibility menu, dialog, popup

UserName

CAS RN 53797-35-6

IUPAC Name: (2R,3S,4R,5R,6R)-5-amino-2-(aminomethyl)-6-{[(1R,2R,3S,4R,6S)-4,6-diamino-2-{[(2S,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]oxy}-3-hydroxycyclohexyl]oxy}oxane-3,4-diol; sulfuric acid
Synonyms: ribostamycin; sulfuric acid
Molecular Weight (g/mol): 552.55
Molecular Formula: C17H36N4O14S
InChi Key: RTCDDYYZMGGHOE-YMSVYGIHSA-N
SMILES: OS(O)(=O)=O.NC[C@H]1O[C@H](O[C@@H]2[C@@H](N)C[C@@H](N)[C@H](O)[C@H]2O[C@@H]2O[C@H](CO)[C@@H](O)[C@H]2O)[C@H](N)[C@@H](O)[C@@H]1O
Physical Form 
  • (1)
  • (1)
Quantity 
  • (2)
Color 
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results