missing translation for 'onlineSavingsMsg'
Learn More

Ribostamycin sulfate, 630^mg/mg

Catalog No. p-7076646
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6615503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ6615503 Supplier Thermo Scientific Chemicals Supplier No. J6615503

Used as a broad spectrum antibiotic targeting both Gram-positive and Gram-negative bacteria. Ribostamycin is a broad-spectrum antimicrobial isolated from Streptomyces ribosifidicus. It is used in pharmacokinetic studies. | CAS: 53797-35-6 | C17H36N4O14S | 552.55 g/mol

Used in pharmacokinetic studies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 53797-35-6
Molecular Formula C17H36N4O14S
Molecular Weight (g/mol) 552.55
MDL Number MFCD23380549
InChI Key RTCDDYYZMGGHOE-YMSVYGIHSA-N
Synonym ribostamycin sulfate
PubChem CID 53486170
IUPAC Name (2R,3S,4R,5R,6R)-5-amino-2-(aminomethyl)-6-{[(1R,2R,3S,4R,6S)-4,6-diamino-2-{[(2S,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]oxy}-3-hydroxycyclohexyl]oxy}oxane-3,4-diol; sulfuric acid
SMILES OS(O)(=O)=O.NC[C@H]1O[C@H](O[C@@H]2[C@@H](N)C[C@@H](N)[C@H](O)[C@H]2O[C@@H]2O[C@H](CO)[C@@H](O)[C@H]2O)[C@H](N)[C@@H](O)[C@@H]1O

Specifications

Melting Point 175°C to 180°C
Flash Point >110°C (230°F)
Packaging Plastic bottle
Quantity 1 g
Solubility Information Soluble in water
Formula Weight 552.55
Physical Form Powder
Chemical Name or Material Ribostamycin sulfate
Hazard Category H312+H332-H360
Precautionary Statement P201-P202-P261-P271-P281-P302+P352-P304+P340-P308+P313-P312-P363-P501c
EINECSNumber 258-783-1
RTECSNumber WK2300000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only