missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RiboMinus™ Eukaryote System v2

Catalog No. A15026
Encompass
Change view
Click to view available options
Quantity:
12 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A15026 12 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A15026 Supplier Invitrogen™ Supplier No. A15026
Get Up to $3,000 of qPCR Products with Purchase
View all promotions

The RiboMinus™ Eukaryote System v2 provides truly complete transcriptome isolation for next generation sequencing analysis by offering selective depletion of up to 99.9% of the cytoplasmic (5S, 5.8S, 18S, and 28S) and mitochondrial (12S and 16S) ribosomal RNA from total RNA.

The RiboMinus™ Eukaryote System v2 provides truly complete transcriptome isolation for next generation sequencing analysis by offering selective depletion of up to 99.9% of the cytoplasmic (5S, 5.8S, 18S, and 28S) and mitochondrial (12S and 16S) ribosomal RNA from total RNA. This selection system contrasts with poly-A and cap binding methods, which introduce bias and select for only part of the transcriptome.

Features of the RiboMinus™ Eukaryote System v2 include:
• Magnetic bead-based purification for rapid, seamless purification
• A 1-hour workflow
• An input range of 1–5 μg

The RiboMinus™ Eukaryote System v2 improves upon the previous system (RiboMinus™ Eukaryote Kit for RNA-Seq) by removing significantly more rRNA (both cytoplasmic and mitochondrial) from the total RNA. When used with our library construction kits for sequencing, the RiboMinus™ Eukaryote System v2 provides maximum mapping to reference database with minimum bias, maintaining relative gene expression levels of even smaller RNA species. The resulting total RNA after depletion contains minimum contaminating rRNA with reduced variability.

The RiboMinus Eukaryote System v2 is designed for the removal of human, mouse, and rat rRNA. A magnetic bead-based purification module is included for purification of the whole transcriptome without altering the expression levels of small or non-coding RNA. For column-based purification, please see the RiboMinus™ Eukaryote Kit v2.The RiboMinus Eukaryote System v2 has an RNA input range of 1–5 μg. For lower inputs (down to 100 ng), the Low Input RiboMinus™ Eukaryote System v2 is recommended.

The RiboMinus™ Eukaryote System v2 contains sufficient reagents to perform 12 ribosomal RNA depletions.

Specifications

Content And Storage • RiboMinus™ Eukaryote Probe Mix v2, store at -20°C
• RiboMinus™ Magnetic Beads, store at 4°C
• 2X Hybridization Buffer, store at room temperature
• Binding Solution Concentrate, store at room temperature
• Wash Solution Concentrate, store at room temperature
• Nucleic Acid Binding Beads, store at 4°C
Species Human, Mouse, Rat
Final Product Type Transcriptome RNA
For Use With (Equipment) Ion PGM™ System, Ion Proton™ System, Illumina™ Genome Analyzer II (GA II)
No. of Reactions 12 reactions
Product Line RiboMinus
Quantity 12 reactions
Sequencing Type Transcriptome Sequencing
Starting Material Amount 1 - 5 μg total RNA
Workflow Step Sample Enrichment
I used the RiboMinus Eukaryote System v2 for total transcriptome isolation but I am seeing genomic DNA contamination. Why is this?

This is most likely due to contamination of the total RNA sample with genomic DNA. Before the RiboMinus Kit procedure, we recommend treating the total RNA sample with DNase I to remove any genomic DNA contamination.

For Research Use Only. Not for use in diagnostic procedures.