missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ RiboJuice™ mRNA Transfection Kit

Catalog No. TR1013
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
TR1013 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. TR1013 Supplier MilliporeSigma™ Supplier No. TR1013

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Delivers larger RNA species into mammalian cells with high efficiencies low toxicities.

  • Increases IPS reprogramming efficiencies when used with Simplicon RNA reprogramming technology
  • Increased iPSC Reprogramming efficiencies: 5-fold increase in IPS cell generation using RNA reprogramming methods
  • High efficiency RNA delivery: Achieve RNA delivery in large population of cells to ensure experimental success
  • Low cellular toxicity: Maintain cell density reduce experimental biases
  • Serum compatibility: Perform transfections in presence of serum which eliminates need for media change maintains cellular health
  • Animal free format

Specifications

Content And Storage 2°C to 8°C
Includes Contains two components. RiboJuice™ mRNA Transfection Reagent: One vial containing 0.4 mL of transfection reagent. Store at 2-8°C.  RiboJuice™ mRNA Boost Reagent: One vial containing 0.4 mL of boost reagent. Store at 2-8°C.
Quantity 1 Kit