missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Riboflavin Tetrabutyrate 98.0+%, TCI America™
Supplier: TCI America R005525G
Specifications
| Riboflavin Tetrabutyrate | |
| 149°C | |
| MFCD00059691 | |
| Vitamin B2 Tetrabutyrate | |
| CCCC(=O)OCC(C(C(CN1C2=C(C=C(C(=C2)C)C)N=C3C1=NC(=O)NC3=O)OC(=O)CCC)OC(=O)CCC)OC(=O)CCC | |
| 656.733 | |
| 656.73 | |
| Crystalline Powder |
| 752-56-7 | |
| C33H44N4O10 | |
| 25 g | |
| MJNIWUJSIGSWKK-FSGGQHMVSA-N | |
| [(2S,3R,4R)-2,3,4-tri(butanoyloxy)-5-(7,8-dimethyl-2,4-dioxobenzo[g]pteridin-10-yl)pentyl] butanoate | |
| 18640873 | |
| ≥98.0% (HPLC) |
Chemical Identifiers
| 752-56-7 | |
| 656.733 | |
| MJNIWUJSIGSWKK-FSGGQHMVSA-N | |
| 18640873 | |
| CCCC(=O)OCC(C(C(CN1C2=C(C=C(C(=C2)C)C)N=C3C1=NC(=O)NC3=O)OC(=O)CCC)OC(=O)CCC)OC(=O)CCC |
| C33H44N4O10 | |
| MFCD00059691 | |
| Vitamin B2 Tetrabutyrate | |
| [(2S,3R,4R)-2,3,4-tri(butanoyloxy)-5-(7,8-dimethyl-2,4-dioxobenzo[g]pteridin-10-yl)pentyl] butanoate |
Safety and Handling
RTECSNumber : VJ1755000
TSCA : No