accessibility menu, dialog, popup

UserName

H3COO(R)OOH3C(S)OOCH3(S)NCH3CH3NOHNONOOCH3

CAS RN 752-56-7

IUPAC Name: (2R,3S,4S)-1,2,4-tris(butanoyloxy)-5-{7,8-dimethyl-2,4-dioxo-2H,3H,4H,10H-benzo[g]pteridin-10-yl}pentan-3-yl butanoate
Synonyms: riboflavin tetrabutyrate
Molecular Weight (g/mol): 656.73
Molecular Formula: C33H44N4O10
InChi Key: MJNIWUJSIGSWKK-BBANNHEPSA-N
SMILES: CCCC(=O)OC[C@@H](OC(=O)CCC)[C@@H](OC(=O)CCC)[C@H](CN1C2=CC(C)=C(C)C=C2N=C2C(=O)NC(=O)N=C12)OC(=O)CCC
Grade 
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results