missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Riboflavin Impurity A Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™
Specifications
Specifications
| Purity Grade Notes | These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available. |
| Linear Formula | C13H12N4O2 |
| Synonym | Riboflavin Impurity A; 7,8,10-trimethylbenzo[g]pteridine-2,4(3 H,10 H)-dione (lumiflavine) |
| Type | Certified Reference Material |
| Quantity | 10 mg |
| Formula Weight | 256.26 |
| Grade | Certified Reference Material |