missing translation for 'onlineSavingsMsg'
Learn More

Riboflavin (B2), MilliporeSigma™ Supelco™

Catalog No. 111015656
Change view
Click to view available options
Quantity:
1000 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015656 1000 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015656 Supplier Merck Emd Millipore Supplier No. 47861
Only null left
Cleaning Products, Cosmetics, Food and Beverages, Personal Care and Pharmaceutical (small molecule) Vitamins, Nutraceuticals and Natural Products

Specifications

Packaging Ampule
CAS 83-88-5
Physical Form Neat
Linear Formula C17H20N4O6
MDL Number MFCD00005022
Melting Point 290°C
Synonym (-)-Riboflavin; Lactoflavin; Vitamin B2; Vitamin G
Type Food Additive Standard
Quantity 1000 mg
Formula Weight 376.36
Grade Analytical Standard
Show More Show Less