Learn More
Medisca Riboflavin-5-Phosphate Sodium, USP (Dietary Supplement Grade)
Description
Riboflavin-5-Phosphate Sodium, USP is a fine, orange-yellow, crystalline powder with a slight odor. It is hygroscopic.
Dietary Supplement Grade refers to a nutritional supplement that is manufactured by an FDA-registered dietary supplement manufacturer or manufacturer with other certifications and registrations, based on applicable laws and regulations. Qualified manufacturers follow their applicable standards of practice, which are less stringent than those for FDA-registered drug manufacturers. The customer is responsible for ensuring suitability of the product for the intended use and acknowledges that further testing may be required.
Alternative name(s): Vitamin B2 phosphate
Specifications
Specifications
| Chemical Name or Material | Riboflavin-5-Phosphate Sodium, USP (Dietary Supplement Grade) |
| Packaging | Jar, 1500mL, White, HDPE, Round, 89-400, TE Pano Neck |
| CAS | 130-40-5 |
| Type | Drug Standard |
| Molecular Weight (g/mol) | 514.3576 |
| Quantity | 500 g |
| Percent Purity | 73 to 79 % (Riboflavin) |
| Grade | USP |