Learn More
Riboflavin-5'-phosphate sodium salt dihydrate
CAS: 6184-17-4 | C17H24N4NaO11P | 514.36 g/mol
$95.33 - $342.47
Chemical Identifiers
| CAS | 6184-17-4 |
|---|---|
| Molecular Formula | C17H24N4NaO11P |
| Molecular Weight (g/mol) | 514.36 |
| MDL Number | MFCD00150993 |
| InChI Key | LQVGUKOCMOKKJU-FTNKLFHDNA-M |
| Synonym | riboflavin 5'-monophosphate sodium salt dihydrate |
| PubChem CID | 131856174 |
| IUPAC Name | [(2R,3S,4S)-5-(7,8-dimethyl-2,4-dioxobenzo[g]pteridin-10-yl)-2,3,4-trihydroxypentyl] dihydrogen phosphate;sodium;dihydrate |
| SMILES | O.O.[Na+].CC1=C(C)C=C2N(C[C@H](O)[C@H](O)[C@H](O)COP(O)([O-])=O)C3=NC(=O)NC(=O)C3=NC2=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1454509
|
Thermo Scientific Chemicals
A1454509 |
10 g |
Each for $95.33
|
|
|||||
|
AAA1454518
|
Thermo Scientific Chemicals
A1454518 |
50 g |
Each for $342.47
|
|
|||||
Description
Riboflavin 5?-monophosphate (FMN) is used as a prosthetic group by oxidoreductase (one and two electron transfer) enzymes such as NADH dehydrogenase, nitric oxide synthase, and nitrilotriacetate monooxygenase. Riboflavin 5?-monophosphate is used as a substrate to study the specificity and kinetics of FMN phosphohydrolases and to study molecular sensing based gene regulation by riboswitches. It is a photoinitiator of polymerization for PAGE gels.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsRiboflavin 5′-monophosphate (FMN) is used as a prosthetic group by oxidoreductase (one and two electron transfer) enzymes such as NADH dehydrogenase, nitric oxide synthase, and nitrilotriacetate monooxygenase. Riboflavin 5′-monophosphate is used as a substrate to study the specificity and kinetics of FMN phosphohydrolases and to study molecular sensing based gene regulation by riboswitches. It is a photoinitiator of polymerization for PAGE gels.
Solubility
Soluble in water 50 g/L @ 20°C.
Notes
Store in refrigerator. Protect from heat. Store away from oxidizing agents.
Chemical Identifiers
| 6184-17-4 | |
| 514.36 | |
| LQVGUKOCMOKKJU-FTNKLFHDNA-M | |
| 131856174 | |
| O.O.[Na+].CC1=C(C)C=C2N(C[C@H](O)[C@H](O)[C@H](O)COP(O)([O-])=O)C3=NC(=O)NC(=O)C3=NC2=C1 |
| C17H24N4NaO11P | |
| MFCD00150993 | |
| riboflavin 5'-monophosphate sodium salt dihydrate | |
| [(2R,3S,4S)-5-(7,8-dimethyl-2,4-dioxobenzo[g]pteridin-10-yl)-2,3,4-trihydroxypentyl] dihydrogen phosphate;sodium;dihydrate |
Specifications
| 6184-17-4 | |
| C17H24N4NaO11P | |
| 10 g | |
| 14,8201 | |
| Soluble in water 50g/L @ 20°C. | |
| O.O.[Na+].CC1=C(C)C=C2N(C[C@H](O)[C@H](O)[C@H](O)COP(O)([O-])=O)C3=NC(=O)NC(=O)C3=NC2=C1 | |
| 514.36 | |
| 514.36 (478.34 Anhydrous) |
| >300°C | |
| MFCD00150993 | |
| 4106529 | |
| riboflavin 5'-monophosphate sodium salt dihydrate | |
| LQVGUKOCMOKKJU-FTNKLFHDNA-M | |
| [(2R,3S,4S)-5-(7,8-dimethyl-2,4-dioxobenzo[g]pteridin-10-yl)-2,3,4-trihydroxypentyl] dihydrogen phosphate;sodium;dihydrate | |
| 131856174 | |
| Riboflavin-5'-phosphate sodium salt dihydrate |
Safety and Handling
EINECSNumber : 204-988-6
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only