missing translation for 'onlineSavingsMsg'
Learn More

Riboflavin-5'-phosphate sodium salt dihydrate, 73.0% min.

Catalog No. p-7057568
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6694909 10 g
AAJ6694918 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6694909 Supplier Thermo Scientific Chemicals Supplier No. J6694909
Only null left

CAS: 6184-17-4 | C17H24N4NaO11P | 514.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6184-17-4
Molecular Formula C17H24N4NaO11P
Molecular Weight (g/mol) 514.36
MDL Number MFCD00150993
InChI Key LQVGUKOCMOKKJU-FTNKLFHDNA-M
Synonym riboflavin 5'-monophosphate sodium salt dihydrate
PubChem CID 131856174
IUPAC Name [(2R,3S,4S)-5-(7,8-dimethyl-2,4-dioxobenzo[g]pteridin-10-yl)-2,3,4-trihydroxypentyl] dihydrogen phosphate;sodium;dihydrate
SMILES O.O.[Na+].CC1=C(C)C=C2N(C[C@H](O)[C@H](O)[C@H](O)COP(O)([O-])=O)C3=NC(=O)NC(=O)C3=NC2=C1

Specifications

Melting Point >300°C
Color Yellow to Orange
Quantity 10 g
Beilstein 4106529
Merck Index 14,8201
Solubility Information Soluble in water at 50mg/mL
Formula Weight 514.36 (478.34 Anhydrous)
Physical Form Powder
Chemical Name or Material Riboflavin-5'-phosphate sodium salt dihydrate
EINECSNumber 204-988-6
TSCA Yes
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only