missing translation for 'onlineSavingsMsg'
Learn More

Ribavirin 98.0+%, TCI America™

$41.77 - $110.11

Chemical Identifiers

CAS 36791-04-5
Molecular Formula C8H12N4O5
Molecular Weight (g/mol) 244.207
MDL Number MFCD00058564
InChI Key IWUCXVSUMQZMFG-AFCXAGJDSA-N
Synonym ribavirin, tribavirin, virazole, rebetol, ribavirine, copegus, vilona, ribamide, ribamidil, ribasphere
PubChem CID 37542
ChEBI CHEBI:63580
IUPAC Name 1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2,4-triazole-3-carboxamide
SMILES C1=NC(=NN1C2C(C(C(O2)CO)O)O)C(=O)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
R0077100MG
View Documents Promotion Details
N/A 100 mg
Each for $41.77
Only null left
 
R0077500MG
View Documents Promotion Details
N/A 500 mg
Each for $110.11
Only null left
 
Description

Description

Specifications

Chemical Identifiers

36791-04-5
244.207
IWUCXVSUMQZMFG-AFCXAGJDSA-N
37542
1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2,4-triazole-3-carboxamide
C8H12N4O5
MFCD00058564
ribavirin, tribavirin, virazole, rebetol, ribavirine, copegus, vilona, ribamide, ribamidil, ribasphere
CHEBI:63580
C1=NC(=NN1C2C(C(C(O2)CO)O)O)C(=O)N

Specifications

36791-04-5
C8H12N4O5
ribavirin, tribavirin, virazole, rebetol, ribavirine, copegus, vilona, ribamide, ribamidil, ribasphere
C1=NC(=NN1C2C(C(C(O2)CO)O)O)C(=O)N
244.207
CHEBI:63580
≥98.0% (HPLC,N)
Ribavirin
White
MFCD00058564
IWUCXVSUMQZMFG-AFCXAGJDSA-N
1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2,4-triazole-3-carboxamide
37542
244.21
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (9)-1515

RTECSNumber : XZ4250000

TSCA : No

SDS
Documents

Documents