missing translation for 'onlineSavingsMsg'
Learn More

Ribavirin 98.0+%, TCI America™

Catalog No. R0077100MG
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R0077100MG 100 mg
R0077500MG 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R0077100MG Supplier TCI America Supplier No. R0077100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 36791-04-5
Molecular Formula C8H12N4O5
Molecular Weight (g/mol) 244.207
MDL Number MFCD00058564
InChI Key IWUCXVSUMQZMFG-AFCXAGJDSA-N
Synonym ribavirin, tribavirin, virazole, rebetol, ribavirine, copegus, vilona, ribamide, ribamidil, ribasphere
PubChem CID 37542
ChEBI CHEBI:63580
IUPAC Name 1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2,4-triazole-3-carboxamide
SMILES C1=NC(=NN1C2C(C(C(O2)CO)O)O)C(=O)N

Specifications

Color White
Quantity 100 mg
Formula Weight 244.21
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Ribavirin
EINECSNumber (9)-1515
RTECSNumber XZ4250000
TSCA No