missing translation for 'onlineSavingsMsg'
Learn More

Ribavirin Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111012196
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012196 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012196 Supplier Merck Emd Millipore Supplier No. PHR17001G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Analog of the imidazole nucleotide intermediates in purine nucleotide biosynthesis.

Specifications

CAS 36791-04-5
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C8H12N4O5
MDL Number MFCD00058564
Synonym Ribavirin; 1-beta-D-Ribofuranosyl-1,2,4-triazole-3-carboxamide
Type Certified Reference Material
Quantity 1 g
Formula Weight 244.20
Grade Certified Reference Material