missing translation for 'onlineSavingsMsg'
Learn More

Ribavirin Impurity Standard, MilliporeSigma™ Supelco™

Catalog No. 111002137
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111002137 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111002137 Supplier Merck Emd Millipore Supplier No. BP592

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Analog of the imidazole nucleotide intermediates in purine nucleotide biosynthesis.

Specifications

CAS 36791-04-5
Linear Formula C8H12N4O5
MDL Number MFCD00058564
Synonym Ribavirin; 1-beta-D-Ribofuranosyl-1,2,4-triazole-3-carboxamide
Type Pharmacopeia and Metrological Institutes Standards
Quantity 25 mg
Formula Weight 244.20
Grade Pharmaceutical Primary Standard