missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RIAM Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570306
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570306 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570306 Supplier Invitrogen™ Supplier No. PA570306
Only null left

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 86%.

RIAM appears to function in the signal transduction from Ras activation to actin cytoskeletal remodeling. Suppresses insulin-induced promoter activities through AP1 and SRE. Mediates Rap1-induced adhesion.

Specifications

Antigen RIAM
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene APBB1IP
Gene Accession No. 0
Gene Alias 9930118P07Rik; amyloid beta (A4) precursor protein-binding, family B, member 1 interacting protein; amyloid beta A4 precursor protein-binding family B member 1-interacting protein; amyloid beta precursor protein binding family B member 1 interacting protein; APBB1-interacting protein 1; Apbb1ip; INAG1; Prel1; PREL-1; proline rich EVH1 ligand 1; proline-rich EVH1 ligand 1; Proline-rich protein 48; proline-rich protein 73; Prp48; Rap1-GTP-interacting adapter molecule; Rap1-GTP-interacting adaptor molecule; Rap1-interacting adaptor molecule; RARP1; RARP-1; retinoic acid-responsive proline-rich protein 1; RIAM; RIAMRARP-1; zgc:63968
Gene Symbols APBB1IP
Host Species Rabbit
Immunogen synthetic peptide corresponding to a region of rat RIAM.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 307171
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less