missing translation for 'onlineSavingsMsg'
Learn More

Rhynchophylline 98.0+%, TCI America™

Catalog No. R010525MG
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R010525MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R010525MG Supplier TCI America Supplier No. R010525MG

Chemical Identifiers

CAS 76-66-4
Molecular Formula C22H28N2O4
Molecular Weight (g/mol) 384.476
MDL Number MFCD00221748
InChI Key DAXYUDFNWXHGBE-GXONQVTISA-N
PubChem CID 102004527
IUPAC Name methyl (E)-2-[(6'R,7'S,8'aS)-6'-ethyl-2-oxospiro[1H-indole-3,1'-3,5,6,7,8,8a-hexahydro-2H-indolizine]-7'-yl]-3-methoxyprop-2-enoate
SMILES CCC1CN2CCC3(C2CC1C(=COC)C(=O)OC)C4=CC=CC=C4NC3=O

Specifications

Physical Form Powder/Crystals
Color White/Yellow
Melting Point 206°C
Quantity 25 mg
Formula Weight 384.48
Percent Purity ≥98.0% (HPLC)
Chemical Name or Material Rhynchophylline
RTECSNumber GN1600000
TSCA No
Recommended Storage Refrigerator