missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Rhp51 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA14968
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA14968 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA14968 Supplier Invitrogen™ Supplier No. PA14968

Rabbit Polyclonal Antibody

PA1-4968 detects Rhp51 protein in E. coli samples. PA1-4968 has been successfully used in Western blot, Immunofluorescence, immunocytochemistry and Immunoprecipitation proceedures. It is specific for the ~40 kDa Rhp51 protein. GFP-tagging of Rhp51 protein at either N- or C- terminus inactivates the function of Rhp51, and cannot be used for the study of foci formation. PA1-4968 immunogen is a highly purified recombinant Rhp51 protein expressed in E. coli.

Rhp51 protein of Schizosaccharomyces pombe (fission yeast) is a functional and structural homolog of E. coli RecA protein and the Rad51 proteins of eukaryotes. These proteins play a major role in genetic recombination and recombination repair by mediating strand exchange reaction between homologous DNA strands.

Specifications

Antigen Rhp51
Applications ChIP Assay, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation whole serum with 0.09% sodium azide
Gene Alias RAD51 homolog
Gene Symbols rad51
Host Species Rabbit
Immunogen Full-size purified recombinant Rhp51 protein expressed in E. coli.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Yeast, Bacteria
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less