missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Rhodopsin Monoclonal Antibody (RET-P1)
GREENER_CHOICE

Catalog No. pima511741
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA511741 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA511741 Supplier Invitrogen™ Supplier No. MA511741
Only null left

Mouse Monoclonal Antibody

MA5-11741 targets Rhodopsin in immunofluorescence, immunohistochemistry (paraffin) and Western blot applications and shows reactivity with human, mouse, and rat samples. The MA5-11741 immunogen is membrane preparation from adult rat retina.

Visual pigments such as rhodopsin and porphyropsin are light-absorbing molecules that mediate vision. Rhodopsin consists of an apoprotein, opsin, covalently linked to 11-cis-retinal. This receptor is coupled to the activation of phospholipase C. Porphyropsin consists of opsin covalently linked to 11-cis 3,4-didehydroretinal.

Specifications

Antigen Rhodopsin
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone RET-P1
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4
Gene RHO
Gene Accession No. P08100, P15409, P51489
Gene Alias CSNBAD1; L opsin; Long Wavelength Sensitive opsin; LWS opsin; MGC138309; MGC138311; Noerg1; OPN 2; Opn2; Ops; opsin 2; opsin 2, rod pigment; opsin-2; Red Opsin; retinal rod opsin pigment rh1.1; Rh; RH1; rh1.1; rho; rho protein; rhodopsin; rhodopsin (opsin 2, rod pigment) (retinitis pigmentosa 4, autosomal dominant); Rhodopsin (retinitis pigmentosa 4, autosomal dominant); rhodopsin-like-1.1 rod opsin; Rod Opsin; RP 4; RP4; seven transmembrane light receptor; unknown opsin-like protein; wu:fi06d11; zfo2; zfrho
Gene Symbols RHO
Host Species Mouse
Immunogen Membrane preparation from adult rat retina.
Purification Method Protein G
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 212541, 24717, 6010
Target Species Human, Mouse, Rat
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less