missing translation for 'onlineSavingsMsg'
Learn More

Rhodium(II) Acetate Dimer, TCI America™

Catalog No. p-7135682
Change view
Click to view available options
Quantity:
100 mg
1 g
Catalog No. Quantity
R0069100MG 100 mg
R00691G 1 g
2 options

Catalog No. R0069100MG

Supplier: TCI America R0069100MG

Only null left

Chemical Identifiers

CAS 15956-28-2
Molecular Formula C8H12O8Rh2
Molecular Weight (g/mol) 441.99
MDL Number MFCD00003538
InChI Key VOXKIADTKKPBBP-UHFFFAOYSA-N
Synonym rhodium ii acetate, rhodium diacetate, acetic acid, rhodium 2+ salt, rhodium ii acetate dimer dihydrate, dirhodium tetraacetate, rhodium ii acetate dimer, rhodium acetate, brown, water soluble, diacetoxyrhodium
PubChem CID 122130469
IUPAC Name tetramethyl-2,4,6,8,9,11,12,14-octaoxa-1,5-dirhodatetracyclo[3.3.3.3¹,⁵.0¹,⁵]tetradecane-1,1,5,5-tetrakis(ylium)-3,7,10,13-tetraide
SMILES C[C-]1O[Rh++]234O[C-](C)O[Rh++]2(O1)(O[C-](C)O3)O[C-](C)O4

Specifications

Melting Point 240°C
Quantity 100 mg
Formula Weight 441.99
Physical Form Crystalline Powder
Chemical Name or Material Rhodium(II) Acetate Dimer
RTECSNumber VI9361000
TSCA No