missing translation for 'onlineSavingsMsg'
Learn More

ICP-MS Internal Standard, 100 ppm Rh in 3% HCl, Ricca Chemical

Catalog No. msrh100h100
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MSRH100H100 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MSRH100H100 Supplier Ricca Chemical Company Supplier No. MSRH100H100
Only null left

Chemical Identifiers

CAS 20765-98-4 , 7732-18-5 , 20765-98-4
Molecular Formula Cl3Rh
Molecular Weight (g/mol) 209.26
MDL Number MFCD00149839
InChI Key SONJTKJMTWTJCT-UHFFFAOYSA-K
IUPAC Name rhodium(3+) trichloride
SMILES [Cl-].[Cl-].[Cl-].[Rh+3]

Specifications

Packaging Natural LDPE Bottle
Concentration or Composition Notes 100ppm Rh
CAS Min % 95.0, 3.29, 0.02
CAS Max % 99.0, 3.42, 0.02
Equivalence 1 mL = 0.1 mg Rh
Physical Form Liquid
Color Red
pH <1
Melting Point Approximately 0°C
Boiling Point Approximately 100°C
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Solubility Miscible with water
Solution Type Calibration Standard
Type ICP Single-element Standard Solution
Quantity 100 mL
Grade Laboratory
Chemical Name or Material Rhodium ICP-MS Standard
Show More Show Less