missing translation for 'onlineSavingsMsg'
Learn More

Rhodanine Indicator, 0.02% (w/v), LabChem™

Catalog No. LC222507
Change view
Click to view available options
Quantity:
125 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC222507 125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC222507 Supplier LabChem Supplier No. LC222507

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Laboratory reagent used as an indicator solution.

Chemical Identifiers

CAS 141-84-4 , 7732-18-5
Molecular Formula C3H3NOS2
Molecular Weight (g/mol) 133.183
InChI Key KIWUVOGUEXMXSV-UHFFFAOYSA-N
PubChem CID 1201546
ChEBI CHEBI:8830
IUPAC Name 2-sulfanylidene-1,3-thiazolidin-4-one
SMILES C1C(=O)NC(=S)S1

Specifications

CAS Max % 99.98, 0.02
Identification Passes Test
Packaging Poly Bottle
Specific Gravity 1g/mL
Linear Formula C3H3NOS2
Decomposition Information Nitrogen oxides; Carbon monoxide; Carbon dioxide; Sulfur compounds
Density 1g/mL
Color Yellow
Solubility Information Soluble in water
Quantity 125 mL
Physical Form Liquid
Formula Weight 133.19
Percent Purity 0.02% (w/v)
Grade Indicator
Chemical Name or Material Rhodanine Indicator
Show More Show Less
Hazard Statement GHS H Statement
Solution is not hazardous.
Precautionary Statement GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage Room Temperature