missing translation for 'onlineSavingsMsg'
Learn More

Rhodanine Indicator, 0.02% (w/v), Ricca Chemical

$173.79 - $303.43

Chemical Identifiers

CAS 141-84-4
Molecular Formula C3H3NOS2
Molecular Weight (g/mol) Mixture
InChI Key KIWUVOGUEXMXSV-UHFFFAOYSA-N
PubChem CID 1201546
ChEBI CHEBI:8830
IUPAC Name 2-sulfanylidene-1,3-thiazolidin-4-one
SMILES C1C(=O)NC(=S)S1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
66254
SDS View Documents
Ricca Chemical Company
66254
120 mL Natural Poly Bottle
Each for $173.79
Only null left
 
662516
View Documents
Ricca Chemical Company
662516
500 mL Natural Poly Bottle
Each for $303.43
Only null left
 
Description

Description

Specifications

Chemical Identifiers

141-84-4
Mixture
1201546
2-sulfanylidene-1,3-thiazolidin-4-one
C3H3NOS2
KIWUVOGUEXMXSV-UHFFFAOYSA-N
CHEBI:8830
C1C(=O)NC(=S)S1

Specifications

141-84-4 , 7732-18-5
100.0,0.02
C3H3NOS2
C1C(=O)NC(=S)S1
Mixture
CHEBI:8830
Indicator
Aqueous Solution
120 mL
Rhodanine Indicator
98.0,0.02
0.02% (w/v)
KIWUVOGUEXMXSV-UHFFFAOYSA-N
2-sulfanylidene-1,3-thiazolidin-4-one
1201546
0.02% W/V
Natural Poly Bottle
Golden/Tan
Liquid
Videos
SDS
Documents

Documents