missing translation for 'onlineSavingsMsg'
Learn More

Rhodanine-3-propionic Acid 98.0+%, TCI America™

Catalog No. p-7135676
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R00605G 5 g
R006025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R00605G Supplier TCI America Supplier No. R00605G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7025-19-6
Molecular Formula C6H7NO3S2
Molecular Weight (g/mol) 205.246
MDL Number MFCD00022558
InChI Key SWAHCTPCIUXXTQ-UHFFFAOYSA-N
Synonym N-(2-Carboxyethyl)rhodanine
PubChem CID 81492
IUPAC Name 3-(4-oxo-2-sulfanylidene-1,3-thiazolidin-3-yl)propanoic acid
SMILES C1C(=O)N(C(=S)S1)CCC(=O)O

Specifications

Quantity 5 g
Formula Weight 205.25
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Rhodanine-3-propionic Acid
RTECSNumber XJ6094060
TSCA No