accessibility menu, dialog, popup

UserName

CAS RN 7025-19-6

IUPAC Name: 3-(4-oxo-2-sulfanylidene-1,3-thiazolidin-3-yl)propanoic acid
Synonyms: 3-(4-oxo-2-sulfanylidene-1,3-thiazolidin-3-yl)propanoic acid
Molecular Weight (g/mol): 205.25
Molecular Formula: C6H7NO3S2
InChi Key: SWAHCTPCIUXXTQ-UHFFFAOYSA-N
SMILES: OC(=O)CCN1C(=O)CSC1=S
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results