missing translation for 'onlineSavingsMsg'
Learn More

Rhodanine-3-acetic Acid 98.0+%, TCI America™

Catalog No. p-7135674
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R00545G 5 g
R005425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R00545G Supplier TCI America Supplier No. R00545G
Only null left

Chemical Identifiers

CAS 5718-83-2
Molecular Formula C5H5NO3S2
Molecular Weight (g/mol) 191.22
MDL Number MFCD00005491
InChI Key JGRMXPSUZIYDRR-UHFFFAOYSA-N
Synonym rhodanine-3-acetic acid, rhodanine-n-acetic acid, n-carboxymethylrhodanine, 3-rhodanineacetic acid, 3-thiazolidineacetic acid, 4-oxo-2-thioxo, 3-carboxymethyl rhodanine, n-carboxymethyl rhodanine, 2-4-oxo-2-thioxothiazolidin-3-yl acetic acid, 4-oxo-2-thioxo-3-thiazolidineacetic acid, 4-oxo-2-thioxo-3-thiazolidinylacetic acid
PubChem CID 79793
IUPAC Name 2-(4-oxo-2-sulfanylidene-1,3-thiazolidin-3-yl)acetic acid
SMILES OC(=O)CN1C(=O)CSC1=S

Specifications

Melting Point 147°C
Quantity 5 g
Formula Weight 191.22
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Rhodanine-3-acetic Acid
EINECSNumber (5)-0223
TSCA Yes