accessibility menu, dialog, popup

UserName

CAS RN 5718-83-2

IUPAC Name: 2-(4-oxo-2-sulfanylidene-1,3-thiazolidin-3-yl)acetic acid
Synonyms: (4-oxo-2-sulfanylidene-1,3-thiazolidin-3-yl)acetic acid
Molecular Weight (g/mol): 191.22
Molecular Formula: C5H5NO3S2
InChi Key: JGRMXPSUZIYDRR-UHFFFAOYSA-N
SMILES: OC(=O)CN1C(=O)CSC1=S
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)