missing translation for 'onlineSavingsMsg'
Learn More

Rhodanine-3-acetic acid, 98%

Catalog No. p-7026308
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2224406 5 g
AAB2224414 25 g
AAB2224422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2224406 Supplier Thermo Scientific Chemicals Supplier No. B2224406
Only null left

CAS: 5718-83-2 | C5H5NO3S2 | 191.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5718-83-2
Molecular Formula C5H5NO3S2
Molecular Weight (g/mol) 191.22
MDL Number MFCD00005491
InChI Key JGRMXPSUZIYDRR-UHFFFAOYSA-N
Synonym rhodanine-3-acetic acid, rhodanine-n-acetic acid, n-carboxymethylrhodanine, 3-rhodanineacetic acid, 3-thiazolidineacetic acid, 4-oxo-2-thioxo, 3-carboxymethyl rhodanine, n-carboxymethyl rhodanine, 2-4-oxo-2-thioxothiazolidin-3-yl acetic acid, 4-oxo-2-thioxo-3-thiazolidineacetic acid, 4-oxo-2-thioxo-3-thiazolidinylacetic acid
PubChem CID 79793
IUPAC Name 2-(4-oxo-2-sulfanylidene-1,3-thiazolidin-3-yl)acetic acid
SMILES OC(=O)CN1C(=O)CSC1=S

Specifications

Melting Point 145°C to 148°C
Quantity 5 g
Beilstein 149513
Formula Weight 191.23
Percent Purity 98%
Chemical Name or Material Rhodanine-3-acetic acid
Hazard Category H318
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P280-P305+P351+P338-P310
EINECSNumber 227-220-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only