missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Rhodamine WT, 20% solution in water

CAS: 37299-86-8 | C29H29ClN2Na2O5 | 566.99 g/mol

$150.92 - $841.28

Specifications

Chemical Name or Material Rhodamine WT
CAS 37299-86-8,7732-18-5
Molecular Formula C29H29ClN2Na2O5
Solubility Information Solubility in water: miscible.
InChI Key NRZDMKVYRRMFRR-UHFFFAOYSA-L
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC446971000
View Documents
Thermo Scientific Chemicals
446971000
100 mL
Each for $150.92
Only null left
 
AC446970010
View Documents
Thermo Scientific Chemicals
446970010
1 L
Each for $841.28
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Rhodamine WT
C29H29ClN2Na2O5
NRZDMKVYRRMFRR-UHFFFAOYSA-L
disodium;4-[3-(diethylamino)-6-diethylazaniumylidenexanthen-9-yl]benzene-1,3-dicarboxylate;chloride
37718
Glass bottle
1.1600g/mL
37299-86-8,7732-18-5
Solubility in water: miscible.
CCN(CC)C1=CC2=C(C=C1)C(=C3C=CC(=[N+](CC)CC)C=C3O2)C4=C(C=C(C=C4)C(=O)[O-])C(=O)[O-].[Na+].[Na+].[Cl-]
566.99
566.99
1.16
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only