missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Rhodamine WT, 20% solution in water
CAS: 37299-86-8 | C29H29ClN2Na2O5 | 566.99 g/mol
Supplier: Thermo Scientific Chemicals 446971000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Rhodamine WT | |
| C29H29ClN2Na2O5 | |
| NRZDMKVYRRMFRR-UHFFFAOYSA-L | |
| disodium;4-[3-(diethylamino)-6-diethylazaniumylidenexanthen-9-yl]benzene-1,3-dicarboxylate;chloride | |
| 37718 | |
| Glass bottle | |
| 1.1600g/mL |
| 37299-86-8,7732-18-5 | |
| Solubility in water: miscible. | |
| CCN(CC)C1=CC2=C(C=C1)C(=C3C=CC(=[N+](CC)CC)C=C3O2)C4=C(C=C(C=C4)C(=O)[O-])C(=O)[O-].[Na+].[Na+].[Cl-] | |
| 566.99 | |
| 566.99 | |
| 1.16 | |
| 100 mL |
RUO – Research Use Only