missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Rhodamine Red™-X, Succinimidyl Ester, 5-isomer

Catalog No. r6160
Encompass
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R6160 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R6160 Supplier Invitrogen™ Supplier No. R6160
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rhodamine Red™-X, Succinimidyl Ester, 5-isomer

The amine-reactive Rhodamine Red™-X, succinimidyl ester can be used to create red-fluorescent bioconjugates with excitation/emission maxima ∼560/580 nm. This reactive dye contains an additional seven-atom aminohexanoyl spacer ('X') between the fluorophore and the succinimidyl ester group. This spacer helps to separate the fluorophore from its point of attachment, potentially reducing the interaction of the fluorophore with the biomolecule to which it is conjugated.

Specifications

Quantity 5 mg
Product Type Succinimidyl Ester
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Chemical Reactivity Amine
Shipping Condition Room Temperature
Reactive Moiety Active Ester, Succinimidyl Ester
Label or Dye Rhodamine Red™
Label Type Classic Dyes
Product Line Rhodamine Red
Emission 580
Excitation 560
Show More Show Less
What is the excitation and emission wavelength for rhodamine?

Rhodamine is a generic term for a wide variety of cationic dyes whose fluorescence emission can range from green, orange to red. The table below lists the excitation and emission maxima (nm), as well as molar extinction coefficients (“EC”; cm-1 M-1), for various rhodamine dyes (data derived with dye dissolved in methanol).

Dye Excitation Emission EC
Rhodamine B 568 583 88,000
Rhodamine 123 507 529 101,000
Rhodamine 110 499 521 92,000
Rhodamine 6G 528 551 105,000
XRITC 572 596 92,000

For Research Use Only. Not for use in diagnostic procedures.