missing translation for 'onlineSavingsMsg'
Learn More

Rhodamine 800 98.0+%, TCI America™

Catalog No. r0190200mg
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R0190200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R0190200MG Supplier TCI America Supplier No. R0190200MG
Only null left

Chemical Identifiers

CAS 137993-41-0
Molecular Formula C26H26ClN3O5
Molecular Weight (g/mol) 495.96
MDL Number MFCD00467020
InChI Key HACOCUMLBPNDIN-UHFFFAOYSA-M
PubChem CID 127750
SMILES C1CC2=C3C(=C4C(=C2)C(=C5C=C6CCC[N+]7=C6C(=C5O4)CCC7)C#N)CCCN3C1.[O-]Cl(=O)(=O)=O

Specifications

Color Purple
Quantity 200 mg
UN Number 1479
Formula Weight 495.96
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Rhodamine 800