missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Rhodamine 6G, pure

CAS: 989-38-8 | C28H31ClN2O3 | 479.017 g/mol

Supplier:  Thermo Scientific Chemicals 191410500

Catalog No. AC191410500


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
Specifications

Specifications

Rhodamine 6G
989-38-8
MFCD00012665
Basic Red 1, C.I. 45160
VYXSBFYARXAAKO-UHFFFAOYSA-N
[H+].CCNC1=C(C=C2C(=C1)OC3=CC(=NCC)C(=CC3=C2C4=CC=CC=C4C(=O)OCC)C)C.[Cl-]
479.017
479.01
Glass bottle
Brown to Brown/Red
Fine Crystalline Powder
290.0°C
C28H31ClN2O3
18, V,12, 283
Solubility in water: soluble.
Authentic
ethyl 2-[3-(ethylamino)-6-ethylimino-2,7-dimethylxanthen-9-yl]benzoate;hydron;chloride
51358423
Pure
Complies
50 g

Chemical Identifiers

989-38-8
479.017
VYXSBFYARXAAKO-UHFFFAOYSA-N
51358423
[H+].CCNC1=C(C=C2C(=C1)OC3=CC(=NCC)C(=CC3=C2C4=CC=CC=C4C(=O)OCC)C)C.[Cl-]
C28H31ClN2O3
MFCD00012665
Basic Red 1, C.I. 45160
ethyl 2-[3-(ethylamino)-6-ethylimino-2,7-dimethylxanthen-9-yl]benzoate;hydron;chloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only