missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rhodamine 640 Perchlorate 90.0+%, TCI America™
Specifications
Specifications
| Quantity | 1 g |
| UN Number | 1479 |
| Formula Weight | 591.06 |
| Percent Purity | ≥90.0% (HPLC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Rhodamine 640 Perchlorate |
Safety and Handling
| TSCA | Yes |