missing translation for 'onlineSavingsMsg'
Learn More

Rhodamine 640 Perchlorate 90.0+%, TCI America™

Catalog No. p-7135748
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R0188200MG 200 mg
R01881G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R0188200MG Supplier TCI America Supplier No. R0188200MG
Only null left

Chemical Identifiers

CAS 72102-91-1
Molecular Formula C32H31ClN2O7
Molecular Weight (g/mol) 591.06
MDL Number MFCD00799440
InChI Key DJWWHVKRLDNDJK-UHFFFAOYSA-N
PubChem CID 153490
IUPAC Name 16-(2-carboxyphenyl)-3λ⁴-oxa-9,23-diazaheptacyclo[17.7.1.1⁵,⁹.0²,¹⁷.0⁴,¹⁵.0²³,²⁷.0¹³,²⁸]octacosa-1,3,5(28),13,15,17,19(27)-heptaen-3-ylium perchlorate
SMILES [O-][Cl](=O)(=O)=O.OC(=O)C1=CC=CC=C1C1=C2C=C3CCCN4CCCC(C2=[O+]C2=C5CCCN6CCCC(C=C12)=C56)=C34

Specifications

Quantity 200 mg
UN Number 1479
Formula Weight 591.06
Percent Purity ≥90.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Rhodamine 640 Perchlorate