missing translation for 'onlineSavingsMsg'
Learn More

Rhodamine 19 Perchlorate 98.0+%, TCI America™

Catalog No. p-7135749
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R01911G 1 g
R01915G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R01911G Supplier TCI America Supplier No. R01911G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 62669-66-3
Molecular Formula C26H27ClN2O7
Molecular Weight (g/mol) 514.959
MDL Number MFCD02684062
InChI Key WRJTXSZPMAXPRF-UHFFFAOYSA-N
PubChem CID 45157597
IUPAC Name 2-[3-(ethylamino)-6-ethylimino-2,7-dimethylxanthen-9-yl]benzoic acid;perchloric acid
SMILES CCNC1=C(C=C2C(=C1)OC3=CC(=NCC)C(=CC3=C2C4=CC=CC=C4C(=O)O)C)C.OCl(=O)(=O)=O

Specifications

Quantity 1 g
UN Number 1479
Formula Weight 514.96
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Rhodamine 19 Perchlorate