Learn More
Thermo Scientific™ Rheodyne™ 7010 Sample Injector
Catalog No. 03052240
Change view
Click to view available options
Stator Material:
Stainless Steel
Catalog No. Stator Material
03052240 Stainless Steel
1 options
Catalog No. 03052240 Supplier Thermo Scientific™ Supplier No. CMD7010
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Ensure accurate and precise high-volume HPLC injections with the single-mode Thermo Scientific™ Rheodyne™ 7010 Sample Injector.
Ensure accurate and precise high-volume HPLC injections of 5μL to 20mL samples with the Thermo Scientific™ Rheodyne™ 7010 Sample Injector. This single-mode injector uses the complete filling method to load the sample loop.
Specifications
Mode Single Mode
Volume (Metric) Sample 5 μL to 5 mL
Type Sample Injector
No. of Ports 6 Ports
Stator Material Stainless Steel